siRNA Details
| Gene Symbol: LOC441178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_117092.1 | 11 | ||||
| GenBank mRNA | AK056600.1 | - | 11 | |||
| AL832737.1 | - | 4 | 773 | |||
| BC093745.1 | - | 1 | 280 | 7939590 | ||
| BC111990.1 | - | 1 | 281 | 8327458 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC441178| Gene Symbol: LOC441178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_117092.1 | 11 | ||||
| GenBank mRNA | AK056600.1 | - | 11 | |||
| AL832737.1 | - | 4 | 773 | |||
| BC093745.1 | - | 1 | 280 | 7939590 | ||
| BC111990.1 | - | 1 | 281 | 8327458 | ||