siRNA Details
| Gene Symbol: PLEKHG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029884.1 | NP_001025055.1 | 17 | 5217 | ||
| XM_005267064.3 | XP_005267121.1 | 16 | 7296 | |||
| XM_005267065.3 | XP_005267122.1 | 18 | 5243 | |||
| XM_005267066.2 | XP_005267123.1 | 16 | 6100 | |||
| XM_006715521.1 | XP_006715584.1 | 18 | 5470 | |||
| XM_006715522.3 | XP_006715585.1 | 16 | 7337 | |||
| XM_011535981.2 | XP_011534283.1 | 16 | 8093 | |||
| XM_011535982.2 | XP_011534284.1 | 15 | 8054 | |||
| XM_011535983.2 | XP_011534285.1 | 17 | 6518 | |||
| XM_017011102.1 | XP_016866591.1 | 15 | 6063 | |||
| GenBank mRNA | AB033035.1 | - | 10 | 4090 | ||

PLEKHG1
Genomic Map