siRNA Details
| Gene Symbol: FAM71E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308429.1 | NP_001295358.1 | 5 | 1261 | ||
| NM_138411.2 | NP_612420.1 | 5 | 1213 | |||
| XM_011526405.2 | XP_011524707.2 | 5 | 911 | |||
| XM_011526406.2 | XP_011524708.2 | 5 | 863 | |||
| GenBank mRNA | AK125745.1 | - | 5 | 834 | ||
| AK131032.1 | - | 5 | 720 | |||
| BC004941.1 | - | 5 | 672 | |||
| BC071737.1 | - | 5 | 1213 | 5192796 | ||

FAM71E1
Genomic Map