siRNA Details
| Gene Symbol: ITPRIPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034841.3 | NP_001030013.1 | 1 | 2141 | ||
| NR_028028.1 | 2 | 1625 | ||||
| GenBank mRNA | AK096327.1 | - | 2 | 1625 | ||
| AK097274.1 | - | 1 | 1587 | |||
| BC101749.1 | - | 1 | 1690 | 8069255 | ||
| BC101751.1 | - | 1 | 1690 | 8069257 | ||

ITPRIPL2
Genomic Map