siRNA Details
| Gene Symbol: ITPRIPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034841.3 | NP_001030013.1 | 1 | 555 | ||
| GenBank mRNA | BC101749.1 | - | 1 | 104 | 8069255 | |
| BC101751.1 | - | 1 | 104 | 8069257 | ||
| BX335323.2 | - | 1 | 505 | |||
| BX338638.2 | - | 1 | 497 | |||
| BX382445.2 | - | 1 | 533 | |||

ITPRIPL2
Genomic Map