siRNA Details
| Gene Symbol: PLPP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030059.2 | NP_001025230.1 | 7 | 1429 | ||
| NM_001318166.1 | NP_001305095.1 | 6 | 1304 | |||
| NM_001318167.1 | NP_001305096.1 | 5 | 1240 | |||
| NM_001318168.1 | NP_001305097.1 | 4 | 1149 | |||
| NM_001318169.1 | NP_001305098.1 | 3 | 978 | |||
| NR_134516.1 | 5 | 941 | ||||
| XM_005269592.2 | XP_005269649.1 | 6 | 1258 | |||
| XM_006717685.3 | XP_006717748.1 | 7 | 1098 | |||
| XM_011539444.2 | XP_011537746.1 | 8 | 1156 | |||
| XM_017015821.1 | XP_016871310.1 | 4 | 839 | |||
| XM_017015822.1 | XP_016871311.1 | 4 | 806 | |||
| GenBank mRNA | AK098668.1 | - | 7 | 1012 | ||

PLPP4
Genomic Map