siRNA Details
| Gene Symbol: CCDC153 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC101443.1 | - | 1 | 255 | 40022161 | |
| BC101444.1 | - | 1 | 255 | 40022159 | ||
| BC101986.1 | - | 1 | 255 | 40022155 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC153| Gene Symbol: CCDC153 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC101443.1 | - | 1 | 255 | 40022161 | |
| BC101444.1 | - | 1 | 255 | 40022159 | ||
| BC101986.1 | - | 1 | 255 | 40022155 | ||