siRNA Details
| Gene Symbol: DCAF4L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029955.3 | NP_001025126.2 | 1 | 2581 | ||
| GenBank mRNA | AK122914.1 | - | 1 | 2500 | ||
| BC035027.1 | - | 1 | 2454 | 4828469 | ||
| BC052943.1 | - | 1 | 2522 | 5268527 | ||

DCAF4L1
Genomic Map