siRNA Details
| Gene Symbol: RAB40AL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031834.1 | NP_001027004.1 | 1 | 943 | ||
| GenBank mRNA | BC101169.1 | - | 1 | 943 | 40016064 | |
| BC101170.2 | - | 1 | 943 | 40016065 | ||
| BC101171.1 | - | 1 | 943 | 40016067 | ||
| BC101172.1 | - | 1 | 943 | 40016069 | ||

RAB40AL
Genomic Map