siRNA Details
Gene Symbol: SNAI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178310.3 | NP_840101.1 | 1, 2 | 144 | ||
GenBank mRNA | BC041461.1 | - | 1, 2 | 51 | ||
BC107058.2 | - | 1, 2 | 144 | 40005947 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SNAI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178310.3 | NP_840101.1 | 1, 2 | 144 | ||
GenBank mRNA | BC041461.1 | - | 1, 2 | 51 | ||
BC107058.2 | - | 1, 2 | 144 | 40005947 |