siRNA Details
| Gene Symbol: ZNF181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029997.3 | NP_001025168.2 | 4 | 2567 | ||
| NM_001145665.1 | NP_001139137.1 | 4 | 2564 | |||
| XM_005258849.2 | XP_005258906.1 | 5 | 2335 | |||
| XM_005258850.2 | XP_005258907.1 | 5 | 2332 | |||
| XM_005258851.3 | XP_005258908.1 | 5 | 2323 | |||
| XM_006723183.3 | XP_006723246.1 | 5 | 2589 | |||
| XM_006723184.3 | XP_006723247.1 | 5 | 2320 | |||
| XM_017026739.1 | XP_016882228.1 | 5 | 3077 | |||
| XM_017026740.1 | XP_016882229.1 | 4 | 2197 | |||
| GenBank mRNA | BC043228.1 | - | 3 | 3208 | 5295602 | |

ZNF181
Genomic Map