siRNA Details
| Gene Symbol: NEK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199289.2 | NP_954983.1 | 16 | 1575 | ||
| XM_006719808.3 | XP_006719871.2 | 16 | 1656 | |||
| XM_011535066.2 | XP_011533368.1 | 16 | 1656 | |||
| XM_017020555.1 | XP_016876044.1 | 16 | 1658 | |||
| XM_017020556.1 | XP_016876045.1 | 16 | 1662 | |||
| XM_017020557.1 | XP_016876046.1 | 16 | 1666 | |||
| GenBank mRNA | AK126330.1 | - | 7 | 650 | ||
| BC063885.1 | - | 16 | 1561 | 30383658 | ||

NEK5
Genomic Map