siRNA Details
| Gene Symbol: NEK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199289.2 | NP_954983.1 | 10 | 900 | ||
| XM_006719808.3 | XP_006719871.2 | 10 | 981 | |||
| XM_011535066.2 | XP_011533368.1 | 10 | 981 | |||
| XM_011535068.2 | XP_011533370.1 | 10 | 981 | |||
| XM_017020555.1 | XP_016876044.1 | 10 | 983 | |||
| XM_017020556.1 | XP_016876045.1 | 10 | 987 | |||
| XM_017020557.1 | XP_016876046.1 | 10 | 991 | |||
| GenBank mRNA | BC063885.1 | - | 10 | 886 | 30383658 | |

NEK5
Genomic Map