siRNA Details
| Gene Symbol: ZNF404 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033719.2 | NP_001028891.2 | 2 | 312 | ||
| GenBank mRNA | AK300120.1 | - | 2 | 356 | ||
| AY166785.1 | - | 2 | 256 | |||
| BC101332.1 | - | 2 | 306 | |||
| BC101333.1 | - | 2 | 312 | 40025019 | ||

ZNF404
Genomic Map