siRNA Details
Gene Symbol: ZNF829 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037232.3 | NP_001032309.2 | 6 | 1727 | ||
NM_001171979.1 | NP_001165450.1 | 6 | 1672 | |||
XM_005258876.4 | XP_005258933.1 | 6 | 1550 | |||
XM_011526933.2 | XP_011525235.1 | 6 | 1569 | |||
GenBank mRNA | BC107131.1 | - | 6 | 1612 | 40025539 | |
BC107132.1 | - | 6 | 1618 | 40025541 | ||
BX538288.1 | - | 6 | 1673 |
