siRNA Details
| Gene Symbol: SULT6B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001032377.1 | NP_001027549.1 | 7 | 821 | ||
| GenBank mRNA | BC140797.1 | - | 7 | 821 | 9021665 | |
| BC144573.1 | - | 7 | 810 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SULT6B1| Gene Symbol: SULT6B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001032377.1 | NP_001027549.1 | 7 | 821 | ||
| GenBank mRNA | BC140797.1 | - | 7 | 821 | 9021665 | |
| BC144573.1 | - | 7 | 810 | |||