siRNA Details
| Gene Symbol: EXOSC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020158.3 | NP_064543.3 | 2 | 220 | ||
| GenBank mRNA | AF281134.1 | - | 2 | 220 | ||
| AF285785.1 | - | 2 | 351 | |||
| AK314941.1 | - | 2 | 205 | |||
| BC007742.2 | - | 2 | 203 | 4308795 | ||
| BC107696.1 | - | 2 | 225 | 6453881 | ||

EXOSC5
Genomic Map