siRNA Details
| Gene Symbol: ANKRD20A5P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC022023.1 | - | 3 | 1232 | 4824658 | |
| Gene Symbol: FLJ43315 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC057848.1 | - | 5 | 1384 | ||

ANKRD20A5P
Genomic Map