siRNA Details
Gene Symbol: CYB5RL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031672.2 | NP_001026842.2 | 4, 5 | 677 | ||
GenBank mRNA | AK090456.1 | - | 5 | 768 | ||
BC012781.2 | - | 3, 4 | 461 | |||
BC049198.1 | - | 1, 2 | 185 | |||
BC064419.1 | - | 4, 5 | 650 | |||
BC071735.1 | - | 4, 5 | 626 | |||
CD515193.1 | - | 2, 3 | 423 | |||
DA793739.1 | - | 3, 4 | 528 |
