siRNA Details
| Gene Symbol: NCLN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321463.1 | NP_001308392.1 | 7 | 991 | ||
| NM_020170.3 | NP_064555.2 | 7 | 991 | |||
| GenBank mRNA | AK125002.1 | - | 7 | 1047 | ||
| AK172848.1 | - | 7 | 938 | |||
| AK314044.1 | - | 7 | 926 | |||
| AL136567.1 | - | 7 | 822 | |||
| BC008920.2 | - | 3 | 198 | |||
| BC010064.2 | - | 7 | 907 | 4100399 | ||
| BC013283.2 | - | 7 | 1268 | 3959506 | ||
| BC019091.2 | - | 7 | 907 | 4895124 | ||
| BC025926.1 | - | 7 | 903 | 3509966 | ||
| CR533566.1 | - | 7 | 804 | |||

NCLN
Genomic Map