siRNA Details
| Gene Symbol: LOC613266 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130924.1 | 2 | 1251 | |||
| NR_130925.1 | 2 | 954 | ||||
| GenBank mRNA | AK125594.1 | - | 2 | 954 | ||
| BC043571.1 | - | 2 | 941 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC613266| Gene Symbol: LOC613266 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130924.1 | 2 | 1251 | |||
| NR_130925.1 | 2 | 954 | ||||
| GenBank mRNA | AK125594.1 | - | 2 | 954 | ||
| BC043571.1 | - | 2 | 941 | |||