siRNA Details
| Gene Symbol: FAM229B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033564.2 | NP_001028736.1 | 4 | 654 | ||
| XM_017011174.1 | XP_016866663.1 | 4 | 499 | |||
| XM_017011175.1 | XP_016866664.1 | 3 | 353 | |||
| GenBank mRNA | AL049461.1 | - | 4 | 465 | ||
| BC044912.2 | - | 4 | 521 | 5286266 | ||

FAM229B
Genomic Map