siRNA Details
| Gene Symbol: FAM229B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033564.2 | NP_001028736.1 | 4 | 1600 | ||
| GenBank mRNA | BC044912.2 | - | 4 | 1467 | 5286266 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM229B| Gene Symbol: FAM229B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033564.2 | NP_001028736.1 | 4 | 1600 | ||
| GenBank mRNA | BC044912.2 | - | 4 | 1467 | 5286266 | |