siRNA Details
| Gene Symbol: ZNF704 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033723.2 | NP_001028895.1 | 9 | 4222 | ||
| XM_005251279.4 | XP_005251336.2 | 9 | 4853 | |||
| XM_017013725.1 | XP_016869214.1 | 9 | 4269 | |||
| GenBank mRNA | AK131274.1 | - | 9 | 4222 | ||

ZNF704
Genomic Map