siRNA Details
| Gene Symbol: CLN8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018941.3 | NP_061764.2 | 3 | 6755 | ||
| GenBank mRNA | AK096736.1 | - | 1 | 1449 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CLN8| Gene Symbol: CLN8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018941.3 | NP_061764.2 | 3 | 6755 | ||
| GenBank mRNA | AK096736.1 | - | 1 | 1449 | ||