siRNA Details
| Gene Symbol: RPS19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001022.3 | NP_001013.1 | 5 | 758 | ||
| NM_001321483.1 | NP_001308412.1 | 5 | 435 | |||
| NM_001321484.1 | NP_001308413.1 | 5 | 565 | |||
| NM_001321485.1 | NP_001308414.1 | 5 | 771 | |||
| XM_017027113.1 | XP_016882602.1 | 5 | 753 | |||
| GenBank mRNA | AA165492.1 | - | 3 | 311 | ||
| AK311786.1 | - | 5 | 421 | |||
| BC000023.2 | - | 5 | 455 | 3504083 | ||
| BC007615.2 | - | 4 | 386 | 3354280 | ||
| BC017386.1 | - | 5 | 424 | |||
| BC018616.1 | - | 5 | 758 | 4867036 | ||
| BG398397.1 | - | 5 | 565 | |||
| BU603942.1 | - | 5 | 437 | |||
| CB124389.1 | - | 5 | 464 | |||
| CB125125.1 | - | 5 | 435 | |||
| M81757.1 | - | 5 | 408 | |||

RPS19
Genomic Map