siRNA Details
| Gene Symbol: AP5Z1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014855.2 | NP_055670.1 | 16 | 2088 | ||
| XM_017012864.1 | XP_016868353.1 | 15 | 1915 | |||
| GenBank mRNA | AB007875.1 | - | 16 | 2312 | ||
| AK092068.1 | - | 3 | 1551 | |||
| BC008841.2 | - | 7 | 756 | |||
| BC014240.2 | - | 3 | 279 | |||
| BC037399.1 | - | 13 | 2290 | 5172434 | ||

AP5Z1
Genomic Map