siRNA Details
| Gene Symbol: FOXK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037165.1 | NP_001032242.1 | 9 | 3015 | ||
| XM_011515191.2 | XP_011513493.1 | 11 | 2686 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FOXK1| Gene Symbol: FOXK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037165.1 | NP_001032242.1 | 9 | 3015 | ||
| XM_011515191.2 | XP_011513493.1 | 11 | 2686 | |||