siRNA Details
Gene Symbol: FOXK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037165.1 | NP_001032242.1 | 9 | 4097 | ||
XM_011515191.2 | XP_011513493.1 | 11 | 3768 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FOXK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037165.1 | NP_001032242.1 | 9 | 4097 | ||
XM_011515191.2 | XP_011513493.1 | 11 | 3768 |