siRNA Details
| Gene Symbol: FEM1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020177.2 | NP_064562.1 | 2 | 657 | ||
| XM_005272035.4 | XP_005272092.1 | 2 | 659 | |||
| XM_017009647.1 | XP_016865136.1 | 2 | 682 | |||
| GenBank mRNA | AB058688.1 | - | 1 | 185 | ||
| AF391093.1 | - | 1 | 95 | |||
| AK315803.1 | - | 2 | 435 | |||
| AL365415.1 | - | 2 | 395 | |||
| AY249188.1 | - | 1 | 95 | |||
| BC028369.2 | - | 2 | 659 | 4826273 | ||

FEM1C
Genomic Map