siRNA Details
| Gene Symbol: TMEM75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103558.1 | 1 | 1201 | |||
| GenBank mRNA | AK093424.1 | - | 1 | 1201 | ||
| AX748117.1 | - | 1 | 1201 | |||
| BC137383.1 | - | 1 | 823 | 9021381 | ||
| BC137385.1 | - | 1 | 823 | 9021383 | ||

TMEM75
Genomic Map