siRNA Details
| Gene Symbol: HEPN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037558.2 | NP_001032647.2 | 1 | 560 | ||
| GenBank mRNA | AY275431.1 | - | 1 | 560 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HEPN1| Gene Symbol: HEPN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037558.2 | NP_001032647.2 | 1 | 560 | ||
| GenBank mRNA | AY275431.1 | - | 1 | 560 | ||