siRNA Details
| Gene Symbol: LRRC37A5P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034087.1 | 2 | 631 | |||
| GenBank mRNA | BC031236.1 | - | 2 | 633 | 4838739 | |
| BC040897.1 | - | 2 | 325 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LRRC37A5P| Gene Symbol: LRRC37A5P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034087.1 | 2 | 631 | |||
| GenBank mRNA | BC031236.1 | - | 2 | 633 | 4838739 | |
| BC040897.1 | - | 2 | 325 | |||