siRNA Details
| Gene Symbol: Sult1c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133547.4 | NP_598231.3 | 10 | 2534 | ||
| GenBank mRNA | BC103636.1 | - | 10 | 2534 | 7106590 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sult1c2| Gene Symbol: Sult1c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133547.4 | NP_598231.3 | 10 | 2534 | ||
| GenBank mRNA | BC103636.1 | - | 10 | 2534 | 7106590 | |