siRNA Details
| Gene Symbol: Sult1e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012883.1 | NP_037015.1 | 2 | 229 | ||
| GenBank mRNA | M86758.1 | - | 2 | 229 | ||
| S76489.1 | - | 2 | 128 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sult1e1| Gene Symbol: Sult1e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012883.1 | NP_037015.1 | 2 | 229 | ||
| GenBank mRNA | M86758.1 | - | 2 | 229 | ||
| S76489.1 | - | 2 | 128 | |||