siRNA Details
| Gene Symbol: HMCES | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006109.1 | NP_001006109.1 | 2 | 367 | ||
| NM_020187.2 | NP_064572.2 | 2 | 194 | |||
| XM_005247636.3 | XP_005247693.1 | 2 | 379 | |||
| XM_005247637.3 | XP_005247694.1 | 2 | 229 | |||
| XM_017006877.1 | XP_016862366.1 | 2 | 211 | |||
| GenBank mRNA | AF201934.1 | - | 2 | 207 | ||
| AK025883.1 | - | 1 | 39 | |||
| AK225264.1 | - | 2 | 189 | |||
| AK297501.1 | - | 2 | 192 | |||
| AK309891.1 | - | 2 | 191 | |||
| BC009993.2 | - | 2 | 274 | 4130099 | ||
| BC010125.2 | - | 2 | 157 | 3945258 | ||
| BC050686.1 | - | 2 | 367 | 6155077 | ||
| BC088363.1 | - | 2 | 191 | 30398247 | ||

HMCES
Genomic Map