siRNA Details
| Gene Symbol: P22k15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199266.2 | NP_954887.1 | 1129 | |||
| GenBank mRNA | BC100632.1 | - | 1129 | 5621817 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

P22k15| Gene Symbol: P22k15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199266.2 | NP_954887.1 | 1129 | |||
| GenBank mRNA | BC100632.1 | - | 1129 | 5621817 | ||