siRNA Details
| Gene Symbol: Ndufa10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199495.2 | NP_955789.2 | 10 | 1945 | ||
| GenBank mRNA | BC061985.1 | - | 10 | 1773 | 5599238 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ndufa10| Gene Symbol: Ndufa10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199495.2 | NP_955789.2 | 10 | 1945 | ||
| GenBank mRNA | BC061985.1 | - | 10 | 1773 | 5599238 | |