siRNA Details
| Gene Symbol: Olr1410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000784.1 | NP_001000784.1 | 269 | |||
| XM_006246372.1 | XP_006246434.1 | 1 | 269 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1410| Gene Symbol: Olr1410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000784.1 | NP_001000784.1 | 269 | |||
| XM_006246372.1 | XP_006246434.1 | 1 | 269 | |||