siRNA Details
| Gene Symbol: C21orf62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162495.2 | NP_001155967.2 | 4 | 487 | ||
| NM_001162496.2 | NP_001155968.2 | 2 | 331 | |||
| NM_019596.5 | NP_062542.5 | 3 | 412 | |||
| GenBank mRNA | AF231922.1 | - | 3 | 318 | ||
| AK290983.1 | - | 3 | 355 | |||
| BP214434.1 | - | 3 | 372 | |||
| CN428551.1 | - | 3 | 358 | |||
| DQ656042.2 | - | 3 | 412 | |||

C21orf62
Genomic Map