siRNA Details
| Gene Symbol: Olr1562 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000047.1 | NP_001000047.1 | 68 | |||
| Gene Symbol: Olr1563 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751034.1 | XP_003751082.1 | 1 | 68 | ||

Olr1562
Genomic Map