siRNA Details
| Gene Symbol: CPXM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001184699.1 | NP_001171628.1 | 3 | 515 | ||
| NM_019609.4 | NP_062555.1 | 3 | 515 | |||
| GenBank mRNA | AK027661.1 | - | 3 | 451 | ||
| AK075527.1 | - | 3 | 490 | |||
| AK315027.1 | - | 3 | 502 | |||
| AL541188.3 | - | 3 | 487 | |||
| BC063430.1 | - | 3 | 492 | 5239864 | ||
| BX366559.2 | - | 3 | 497 | |||
| CN308423.1 | - | 3 | 506 | |||

CPXM1
Genomic Map