siRNA Details
| Gene Symbol: Olr611 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000926.1 | NP_001000926.1 | 899 | |||
| Gene Symbol: LOC100909651 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762004.1 | XP_008760226.1 | 3 | 938 | ||

Olr611
Genomic Map