siRNA Details
| Gene Symbol: IRGC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019612.3 | NP_062558.1 | 2 | 632 | ||
| XM_005259058.3 | XP_005259115.1 | 2 | 726 | |||
| XM_006723283.3 | XP_006723346.1 | 2 | 663 | |||
| XM_011527118.2 | XP_011525420.1 | 3 | 666 | |||
| GenBank mRNA | BC033853.1 | - | 2 | 632 | ||
| BC066939.1 | - | 2 | 608 | 4826064 | ||

IRGC
Genomic Map