siRNA Details
| Gene Symbol: Vom1r34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008947.1 | NP_001008947.1 | 1 | 896 | ||
| GenBank mRNA | AY510347.1 | - | 1 | 896 | ||
| Gene Symbol: Vom1r64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167554.1 | NP_001161026.1 | 1 | 896 | ||

Vom1r34
Genomic Map