siRNA Details
| Gene Symbol: Vom1r15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008960.1 | NP_001008960.1 | 1 | 20 | ||
| GenBank mRNA | AY510361.1 | - | 1 | 20 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vom1r15| Gene Symbol: Vom1r15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008960.1 | NP_001008960.1 | 1 | 20 | ||
| GenBank mRNA | AY510361.1 | - | 1 | 20 | ||