siRNA Details
| Gene Symbol: Ly49i5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009501.2 | NP_001009501.1 | 935 | |||
| GenBank mRNA | AY649836.1 | - | 935 | |||
| AY653729.1 | - | 780 | ||||
| AY846264.1 | - | 785 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ly49i5| Gene Symbol: Ly49i5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009501.2 | NP_001009501.1 | 935 | |||
| GenBank mRNA | AY649836.1 | - | 935 | |||
| AY653729.1 | - | 780 | ||||
| AY846264.1 | - | 785 | ||||