siRNA Details
| Gene Symbol: Slc46a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024968.1 | NP_001020139.1 | 3 | 572 | ||
| XM_006248833.2 | XP_006248895.1 | 3 | 400 | |||
| GenBank mRNA | BC091179.1 | - | 3 | 572 | 7325773 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc46a3| Gene Symbol: Slc46a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024968.1 | NP_001020139.1 | 3 | 572 | ||
| XM_006248833.2 | XP_006248895.1 | 3 | 400 | |||
| GenBank mRNA | BC091179.1 | - | 3 | 572 | 7325773 | |