siRNA Details
| Gene Symbol: Slc7a12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011948.1 | NP_001011948.1 | 1033 | |||
| GenBank mRNA | BC078940.1 | - | 1033 | 7109204 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc7a12| Gene Symbol: Slc7a12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011948.1 | NP_001011948.1 | 1033 | |||
| GenBank mRNA | BC078940.1 | - | 1033 | 7109204 | ||