siRNA Details
| Gene Symbol: Amdhd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024990.2 | NP_001020161.2 | 1470 | |||
| GenBank mRNA | BC091278.1 | - | 1530 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amdhd2| Gene Symbol: Amdhd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024990.2 | NP_001020161.2 | 1470 | |||
| GenBank mRNA | BC091278.1 | - | 1530 | |||